Learn More
Biomatik Corporation Bovine Albumin (ALB) ELISA Kit
Supplier:  Biomatik Corporation EKU12376
Encompass_Preferred
Bovine Albumin (ALB) ELISA Kit for detecting ALB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 370.4-30,000ng/ml. Sensitivity: 161.3ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P02769. Gene ID: 280717. Target Synonyms: Serum albumin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3505