Learn More
Biomatik Corporation Human/Mouse/Rat/Dog Insulin (INS) ELISA Kit
Supplier:  Biomatik Corporation EKU12390
Encompass_Preferred
Human/Mouse/Rat/Dog Insulin (INS) ELISA Kit for detecting INS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus), Dog (Canis familiaris; Canine). Detection Range: 6.17-500pg/ml. Sensitivity: 2.74pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3510