Learn More
Biomatik Corporation Bovine Catalase (CAT) ELISA Kit
Supplier:  Biomatik Corporation EKU12399
Encompass_Preferred
Bovine Catalase (CAT) ELISA Kit for detecting CAT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00432. Gene ID: 531682. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3515