Learn More
Biomatik Corporation Mouse Cluster Of Differentiation 86 (CD86) ELISA Kit
Supplier:  Biomatik Corporation EKU12422
Encompass_Preferred
Mouse Cluster Of Differentiation 86 (CD86) ELISA Kit for detecting CD86 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P42082. Gene ID: 12524. Target Synonyms: LAB7-2; B7-2; B70; FUN-1; CD28LG2; CD28 Antigen Ligand 2,B7-2 Antigen; CTLA-4 counter-receptor B7.2; B-Lymphocyte Activation Antigen B7-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3519