Learn More
Biomatik Corporation Bovine Suppressors Of Cytokine Signaling 3 (SOCS3) ELISA Kit
Supplier:  Biomatik Corporation EKU12447
Encompass_Preferred
Bovine Suppressors Of Cytokine Signaling 3 (SOCS3) ELISA Kit for detecting SOCS3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BEG9. Gene ID: 282081. Target Synonyms: ATOD4; CIS3; Cish3; MGC71791; SOCS-3; SSI-3; SSI3; Cytokine-inducible SH2 protein 3; STAT-induced STAT inhibitor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3522