Learn More
Biomatik Corporation Chicken Thyroid Stimulating Hormone (TSH) ELISA Kit
Supplier:  Biomatik Corporation EKU12481
Encompass_Preferred
Chicken Thyroid Stimulating Hormone (TSH) ELISA Kit for detecting TSH in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 61.7-5,000pg/ml. Sensitivity: 24.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q6ZXC3. Gene ID: 414344. Target Synonyms: Thyrotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3533