Learn More
Biomatik Corporation Human Enolase 3 (ENO3) ELISA Kit
Supplier:  Biomatik Corporation EKU12514
Encompass_Preferred
Human Enolase 3 (ENO3) ELISA Kit for detecting ENO3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13929. Gene ID: 2027. Target Synonyms: MSE; Beta Enolase; Enolase 3; 2-phospho-D-glycerate hydro-lyase; Skeletal muscle enolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3541