Learn More
Biomatik Corporation Human coagulation factor VIII (FVⅢ) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC33179
Encompass_Preferred
The Human coagulation factor VIII (FVⅢ) ELISA Kit allows for quantitative determination of coagulation factor VIII, procoagulant component in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 12.5 ng/mL-800 ng/mL. Sensitivity: 3.12 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P00451. Restriction: For Research Use Only.
Catalog No. 50-149-7778