Learn More
Biomatik Corporation Bovine Plasminogen (Plg) Elisa Kit
Supplier:  Biomatik Corporation EKC31016
Encompass_Preferred
Bovine Plasminogen (Plg) Elisa Kit for detecting Plasminogen in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 62.5-4000ng/ml. Sensitivity: 15.6ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P06868; PLG. Target Synonyms: PLG; Plasminogen; EC 3.4.21.7) [Cleaved into: Plasmin heavy chain A; Activation peptide; Plasmin heavy chain A; short form; Plasmin light chain B]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3552