Learn More
Biomatik Corporation Dog C-C Motif Chemokine 5 (Ccl5) Elisa Kit
Supplier:  Biomatik Corporation EKC31054
Encompass_Preferred
Dog C-C Motif Chemokine 5 (Ccl5) Elisa Kit for detecting Chemokine (C-C Motif) Ligand 5 in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.081ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8HYS0; CCL5. Target Synonyms: CCL5; SCYA5C-C motif chemokine 5; Small-inducible cytokine A5; T-cell-specific protein RANTES. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3557