Learn More
Biomatik Corporation Goat Insulin (Ins) Elisa Kit
Supplier:  Biomatik Corporation EKC31085
Encompass_Preferred
Goat Insulin (Ins) Elisa Kit for detecting Insulin(Ins) in serum, plasma, tissue homogenates. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 10-100uIU/ml. Sensitivity: 2uIU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01319; INS. Target Synonyms: ILPR; IRDN; OTTHUMP00000011162; OTTHUMP00000196038; proinsulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3561