Learn More
Biomatik Corporation Hamster adiponectin, C1Q and collagen domain containing (ADIPOQ)ELISA kit
Supplier:  Biomatik Corporation EKC31108
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mesocricetus auratus (Golden hamster). Detection Range: 0.469 ng/mL-30 ng/mL. Sensitivity: 0.117 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: C9DFG2. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3565