Learn More
Biomatik Corporation Human Alzheimer-Associated Neuronal Thread Protein (Ad7C-Ntp) Elisa Kit
Supplier:  Biomatik Corporation EKC31137
Encompass_Preferred
Human Alzheimer-Associated Neuronal Thread Protein (Ad7C-Ntp) Elisa Kit for detecting Alzheimer-Associated Neuronal Thread Protein (Ad7C-Ntp) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 93.75-6000pg/ml. Sensitivity: 23.43pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3568