Learn More
Biomatik Corporation Human Apolipoprotein A5 (Apo-A5) Elisa Kit
Supplier:  Biomatik Corporation EKC31176
Encompass_Preferred
Human Apolipoprotein A5 (Apo-A5) Elisa Kit for detecting Apolipoprotein A-V in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.312ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q6Q788; APOA5. Target Synonyms: Apo-AV; ApoA-V; Apoa5; APOA5_HUMAN; ApoAV; Apolipoprotein A-V; Apolipoprotein A5; RAP3; Regeneration associated protein 3; Regeneration-associated protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3570