Learn More
Biomatik Corporation Human C-Peptide Elisa Kit
Supplier:  Biomatik Corporation EKC31202
Encompass_Preferred
Human C-Peptide Elisa Kit for detecting C-Peptide in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.5-8ng/ml. Sensitivity: 0.25ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3575