Learn More
Biomatik Corporation Human Glycoprotein 130, Gp130 Elisa Kit
Supplier:  Biomatik Corporation EKC31229
Encompass_Preferred
Human Glycoprotein 130, Gp130 Elisa Kit for detecting Glycoprotein 130, Gp130 in serum, plasma, tissue homogenates, saliva, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 0.938-60ng/ml. Sensitivity: 0.516ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P40189; IL6ST. Target Synonyms: CD130; CD130 antigen; CDw130; gp130; GP130 RAPS; IL 6R beta; IL-6 receptor subunit beta; IL-6R subunit beta; IL-6R-beta; IL-6RB; IL6 ST; IL6RB_HUMAN; IL6ST; Interleukin 6 receptor subunit beta; Interleukin receptor beta chain; Interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; Membrane glycoprotein 130; Membrane glycoprotein gp130; Oncostatin M receptor; Oncostatin M receptor alpha subunit; Oncostatin-M receptor subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3577