Learn More
Biomatik Corporation Human Protein Nov Homolog (Nov) Elisa Kit
Supplier:  Biomatik Corporation EKC31317
Encompass_Preferred
Human Protein Nov Homolog (Nov) Elisa Kit for detecting Nephroblastoma Overexpressed Gene in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P48745; NOV. Target Synonyms: CCN family member 3; CCN3; IBP-9; IGF-binding protein 9; IGFBP-9; IGFBP9; Insulin-like growth factor-binding protein 9; Nephroblastoma overexpressed; Nephroblastoma overexpressed gene protein homolog; Nephroblastoma-overexpressed gene protein homolog; NOV; NOV_HUMAN; NovH; Protein NOV homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3585