Learn More
Biomatik Corporation Human Tissue Inhibitors Of Metalloproteinase 4, Timp-4 Elisa Kit
Supplier:  Biomatik Corporation EKC31337
Encompass_Preferred
Human Tissue Inhibitors Of Metalloproteinase 4, Timp-4 Elisa Kit for detecting Timp Metallopeptidase Inhibitor 4 in serum, plasma, tissue homogenates, human milk. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.005ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q99727; TIMP4. Target Synonyms: Metalloproteinase inhibitor 4; TIMP 4; TIMP metallopeptidase inhibitor 4; TIMP-4; Timp4; TIMP4_HUMAN; Tissue inhibitor of metalloproteinase 4; Tissue inhibitor of metalloproteinases 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3588