Learn More
Biomatik Corporation Human Tumor Necrosis Factor Soluble Receptor I, Tnfsr-Ielisa Kit
Supplier:  Biomatik Corporation EKC31349
Encompass_Preferred
Human Tumor Necrosis Factor Soluble Receptor I, Tnfsr-Ielisa Kit for detecting Tumor Necrosis Factor Receptor Superfamily, Member 1A in serum, plasma, tissue homogenates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 65pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P19438; TNFRSF1A. Target Synonyms: CD120a; FPF; MGC19588; p55; p55-R; p60; TBP1; TBPI; TNF R; TNF R55; TNF-R1; TNF-RI; TNFAR; TNFR-I; TNFR1; TNFR55; TNFR60; TNFRI; TNFRSF1a; TNR1A_HUMAN; Tumor necrosis factor receptor 1; Tumor necrosis factor receptor superfamily; member 1A; Tumor necrosis factor receptor type 1; Tumor necrosis factor receptor type I; Tumor necrosis factor-binding protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3590