Learn More
Biomatik Corporation Mouse Clusterin (Clu) Elisa Kit
Supplier:  Biomatik Corporation EKC31386
Encompass_Preferred
Mouse Clusterin (Clu) Elisa Kit for detecting Clusterin in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.277ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q06890; Clu. Target Synonyms: Clu; Apoj; Msgp-2Clusterin; Apolipoprotein J; Apo-J; Clustrin; Sulfated glycoprotein 2; SGP-2) [Cleaved into: Clusterin beta chain; Clusterin alpha chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3596