Learn More
Biomatik Corporation Rabbit arachidonic Acid(AA) ELISA kit
Supplier:  Biomatik Corporation EKC31465
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 78.125 ng/mL-5000 ng/mL. Sensitivity: 69.748 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3616