Learn More
Biomatik Corporation Rabbit Plasminogen, Plg Elisa Kit
Supplier:  Biomatik Corporation EKC31472
Encompass_Preferred
Rabbit Plasminogen, Plg Elisa Kit for detecting Plasminogen in serum, plasma, tissue homogenates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 62.5-4000ng/ml. Sensitivity: 15.6ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P00747. Target Synonyms: Plasmin; Plasmin heavy chain A; Plasmin light chain B; Plasminogen; PLG; PLMN_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3618