Learn More
Biomatik Corporation Rat Arachidonic Acid (Aa) Elisa Kit
Supplier:  Biomatik Corporation EKC31484
Encompass_Preferred
Rat Arachidonic Acid (Aa) Elisa Kit for detecting Arachidonic Acid(Aa) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.125-200ng/ml. Sensitivity: 2.758ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3622