Learn More
Biomatik Corporation Bovine Anti-Mullerian Hormone (Amh) Elisa Kit
Supplier:  Biomatik Corporation EKC31548
Encompass_Preferred
Bovine Anti-Mullerian Hormone (Amh) Elisa Kit for detecting Anti-Mullerian Hormone in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.5-200ng/ml. Sensitivity: 0.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P03972; AMH. Target Synonyms: AMH; Muellerian-inhibiting factor; Anti-Muellerian hormone; AMH; Muellerian-inhibiting substance; MIS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3628