Learn More
Biomatik Corporation Bovine Beta-Casein (Csn2) Elisa Kit
Supplier:  Biomatik Corporation EKC31557
Encompass_Preferred
Bovine Beta-Casein (Csn2) Elisa Kit for detecting Casein Beta in breast milk. Species Reactivity: Bovine (Bos taurus). Detection Range: 14-900ug/ml. Sensitivity: 7ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P02666; CSN2. Target Synonyms: CSN2Beta-casein [Cleaved into: Casoparan; Antioxidant peptide; Casohypotensin]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3630