Learn More
Biomatik Corporation Human Coenzyme Q10 (CoQ10) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC33185
Encompass_Preferred
The Human Coenzyme Q10 (CoQ10) ELISA Kit allows for quantitative determination of Coenzyme Q10 (CoQ10) in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 3.12 ng/mL-50 ng/mL. Sensitivity: 1.56 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7781