Learn More
Biomatik Corporation Bovine Inhibin Beta A Chain (Inhba) Elisa Kit
Supplier:  Biomatik Corporation EKC31663
Encompass_Preferred
Bovine Inhibin Beta A Chain (Inhba) Elisa Kit for detecting Inhibin, Beta A in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 15-600pg/ml. Sensitivity: 15pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P07995; INHBA. Target Synonyms: INHBAInhibin beta A chain; Activin beta-A chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3641