Learn More
Biomatik Corporation Bovine Major Prion Protein (Prnp) Elisa Kit
Supplier:  Biomatik Corporation EKC31723
Encompass_Preferred
Bovine Major Prion Protein (Prnp) Elisa Kit for detecting Major Prion Protein(Prnp) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.01-10ng/ml. Sensitivity: 0.01ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10279; PRNP. Target Synonyms: PRNP; PRP; Major prion protein; PrP; Major scrapie-associated fibril protein 1; CD antigen CD230. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3647