Learn More
Biomatik Corporation Bovine Pregnancy Specific Protein B (Pspb) Elisa Kit
Supplier:  Biomatik Corporation EKC31763
Encompass_Preferred
Bovine Pregnancy Specific Protein B (Pspb) Elisa Kit for detecting Pregnancy-Associated Glycoprotein 1 in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.06-24ng/ml. Sensitivity: 0.06ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q29432. Target Synonyms: Pregnancy-associated glycoprotein 1; PAG 1; EC 3.4.23.-; Pregnancy-specific protein B; PSP-B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3652