Learn More
Biomatik Corporation Bovine Serum Amyloid A, Saa Elisa Kit
Supplier:  Biomatik Corporation EKC31786
Encompass_Preferred
Bovine Serum Amyloid A, Saa Elisa Kit for detecting Serum Amyloid A1 in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.1-20ug/ml. Sensitivity: 0.05ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P35541; SAA1. Target Synonyms: SAA1; SAA; Serum amyloid A protein; SAA) [Cleaved into: Amyloid protein A; Amyloid fibril protein AA)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3655