Learn More
Biomatik Corporation Bovine Trypsin Elisa Kit
Supplier:  Biomatik Corporation EKC31815
Encompass_Preferred
Bovine Trypsin Elisa Kit for detecting Trypsin in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3657