Learn More
Biomatik Corporation Bovine Vascular Endothelial Growth Factor A (Vegfa) Elisa Kit
Supplier:  Biomatik Corporation EKC31822
Encompass_Preferred
Bovine Vascular Endothelial Growth Factor A (Vegfa) Elisa Kit for detecting Vascular Endothelial Growth Factor A in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 47-3000pg/ml. Sensitivity: 11.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P15691; VEGFA. Target Synonyms: VEGFA; VEGFVascular endothelial growth factor A; VEGF-A; Vascular permeability factor; VPF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3659