Learn More
Biomatik Corporation Canine Fibroblast Growth Factor 23 (Fgf23) Elisa Kit
Supplier:  Biomatik Corporation EKC31849
Encompass_Preferred
Canine Fibroblast Growth Factor 23 (Fgf23) Elisa Kit for detecting Fibroblast Growth Factor 23 (Fgf23) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: ADHR; HPDR2; HYPF; PHPTC; tumor-derived hypophosphatemia inducing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3665