Learn More
Biomatik Corporation Canine Parathyroid Hormone (Pth) Elisa Kit
Supplier:  Biomatik Corporation EKC31872
Encompass_Preferred
Canine Parathyroid Hormone (Pth) Elisa Kit for detecting Parathyroid Hormone(Pth) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P52212; PTH. Target Synonyms: PTH; Parathyroid hormone; PTH; Parathyrin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3672