Learn More
Biomatik Corporation Canine Relaxin (Rln) Elisa Kit
Supplier:  Biomatik Corporation EKC31878
Encompass_Preferred
Canine Relaxin (Rln) Elisa Kit for detecting Relaxin(Rln) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9TRM8; RLN. Target Synonyms: RLN; Prorelaxin [Cleaved into: Relaxin B chain; Relaxin A chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3676