Learn More
Biomatik Corporation Chicken Growth Hormone, Gh Elisa Kit
Supplier:  Biomatik Corporation EKC31930
Encompass_Preferred
Chicken Growth Hormone, Gh Elisa Kit for detecting Growth Hormone, Gh in serum, plasma, tissue homogenates. Species Reactivity: Chicken (Gallus). Detection Range: 625-10000pg/ml. Sensitivity: 625pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P08998; GH. Target Synonyms: GH; Somatotropin; Growth hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3682