Learn More
Biomatik Corporation Chicken Transforming Growth Factor Β1, Tgf-Β1 Elisa Kit
Supplier:  Biomatik Corporation EKC31991
Encompass_Preferred
Chicken Transforming Growth Factor Β1, Tgf-Β1 Elisa Kit for detecting Transforming Growth Factor, Beta 1 in serum, plasma, tissue homogenates. Species Reactivity: Chicken (Gallus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P09531; TGFB1. Target Synonyms: TGFB1; Transforming growth factor beta-1 proprotein [Cleaved into: Latency-associated peptide; LAP); Transforming growth factor beta-1; TGF-beta-1)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3691