Learn More
Biomatik Corporation Dog Intercellular Adhesion Molecule 1, Icam-1/Cd54 Elisa Kit
Supplier:  Biomatik Corporation EKC32062
Encompass_Preferred
Dog Intercellular Adhesion Molecule 1, Icam-1/Cd54 Elisa Kit for detecting Intercellular Adhesion Molecule 1 in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 25-1600ng/ml. Sensitivity: 6.25ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P33729; ICAM1. Target Synonyms: ICAM1; Intercellular adhesion molecule 1; ICAM-1; CD antigen CD54; Fragment. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3700