Learn More
Biomatik Corporation Dog Orexin (Hcrt) Elisa Kit
Supplier:  Biomatik Corporation EKC32098
Encompass_Preferred
Dog Orexin (Hcrt) Elisa Kit for detecting Hypocretin (Orexin) Neuropeptide Precursor in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 1.87-120pg/ml. Sensitivity: 0.47pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9GLF6; HCRT. Target Synonyms: HCRT; OX; PPOX; Orexin; Hypocretin; Hcrt) [Cleaved into: Orexin-A; Hypocretin-1; Hcrt1); Orexin-B; Hypocretin-2; Hcrt2)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3705