Learn More
Biomatik Corporation Dog S100 Calcium Binding Protein B (S100B) Elisa Kit
Supplier:  Biomatik Corporation EKC32115
Encompass_Preferred
Dog S100 Calcium Binding Protein B (S100B) Elisa Kit for detecting S100 Calcium Binding Protein B (S100B) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 39-2500pg/ml. Sensitivity: 9.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: E2QTP3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3707