Learn More
Biomatik Corporation Duck Interferon Γ , Ifn-Γ Elisa Kit
Supplier:  Biomatik Corporation EKC32133
Encompass_Preferred
Duck Interferon Γ , Ifn-Γ Elisa Kit for detecting Interferon Γ , Ifn-Γ in serum, plasma, tissue homogenates. Species Reactivity: Duck. Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9YGB9; IFNG. Target Synonyms: IFNG; Interferon gamma; IFN-gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3710