Learn More
Biomatik Corporation Mouse Tryptophan Hydroxylase 2 (TPH2) ELISA Kit
Supplier:  Biomatik Corporation EKU07931
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8CGV2. Target Synonyms: NTPH; Neuronal tryptophan hydroxylase; Tryptophan 5-monooxygenase 2. Restriction: For Research Use Only.
Catalog No. 50-229-2972