Learn More
Biomatik Corporation Mouse Tubulin Polymerization Promoting Protein (TPPP) ELISA Kit
Supplier:  Biomatik Corporation EKU07939
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.053ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: TPPP1; p25alpha; p25; TPPP/p25; Brain Specific Protein p25 Alpha; 25 kDa brain-specific protein. Restriction: For Research Use Only.
Catalog No. 50-229-2974