Learn More
Biomatik Corporation Human Tumor Necrosis Factor Alpha Induced Protein 2 (TNFaIP2) ELISA Kit
Supplier:  Biomatik Corporation EKU07953
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.121ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: Primary response gene B94 protein. Restriction: For Research Use Only.
Catalog No. 50-229-2976