Learn More
Biomatik Corporation Rat Tumor Necrosis Factor Receptor 1 (TNFR1) ELISA Kit
Supplier:  Biomatik Corporation EKU07974
Encompass_Preferred
Rat Tumor Necrosis Factor Receptor 1 (TNFR1) ELISA Kit for detecting TNFR1 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P22934. Gene ID: 25625. Target Synonyms: CD120A; P55; TNFRSF1A; TBP1; FPF; TNF-R; TNF-R-I; TNF-R55; TNFAR; TNFR55; TNFR60; P55-R; P60; Tumor necrosis factor receptor 1; Tumor necrosis factor-binding protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2981