Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor Superfamily, Member 1B (TNFRSF1B) ELISA Kit
Supplier:  Biomatik Corporation EKU07975
Encompass_Preferred
Human Tumor Necrosis Factor Receptor Superfamily, Member 1B (TNFRSF1B) ELISA Kit for detecting TNFRSF1B in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20333. Gene ID: 7133. Target Synonyms: CD120B; P75; TBPII; TNF-R-II; TNF-R75; TNFBR; TNFR2; TNFR80; P75tnfr; Tumor necrosis factor receptor 2; p80 TNF-alpha receptor; Etanercept. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2982