Learn More
Biomatik Corporation Horse Vascular Endothelial Growth Factor A (VEGFA) ELISA Kit
Supplier:  Biomatik Corporation EKU08088
Encompass_Preferred
Horse Vascular Endothelial Growth Factor A (VEGFA) ELISA Kit for detecting VEGFA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9GKR0. Gene ID: 100033839. Target Synonyms: VEGF; VEGF-A; VPF; Vascular Endothelial Cell Growth Factor A; Vascular Permeability Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2993