Learn More
Biomatik Corporation Mouse V-Fos FBJ Murine Osteosarcoma Viral Oncogene Homolog (FOS) ELISA Kit
Supplier:  Biomatik Corporation EKU08137
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.056ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P01101. Target Synonyms: Cfos; C-Fos; G0S7; Proto-oncogene c-Fos; G0/G1 Switch Regulatory Protein 7; Cellular oncogene fos. Restriction: For Research Use Only.
Catalog No. 50-229-3000