Learn More
Biomatik Corporation Hamster C-reactive protein(CRP) ELISA Kit
Supplier:  Biomatik Corporation EKC32290
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mesocricetus auratus (Golden hamster). Detection Range: 12.5 ng/mL-800 ng/mL. Sensitivity: 3.12 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P49262. Target Synonyms: MGC149895, MGC88244, PTX1,. Restriction: For Research Use Only.
Catalog No. 50-229-3730