Learn More
Biomatik Corporation Horse Hypoxia Inducible Factor 1, Alpha Subunit (Basic Helix-Loop-Helix Transcription Factor) (Hif1A) Elisa Kit
Supplier:  Biomatik Corporation EKC32334
Encompass_Preferred
Horse Hypoxia Inducible Factor 1, Alpha Subunit (Basic Helix-Loop-Helix Transcription Factor) (Hif1A) Elisa Kit for detecting Hypoxia Inducible Factor 1, Alpha Subunit (Basic Helix-Loop-Helix Transcription Factor) (Hif1A) in serum, plasma, tissue homogenates. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: F6VG41. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3741